ICP Calibration Standard 15, (10X) VeriSpec®
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
15-component standard in 5% nitric acid
50 ppm: Al, As, Ba, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se Sr, Zn; 500 ppm K
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.