Mercury Standard, for AAS, VeriSpec®, Ricca Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
- NIST traceable
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Caution: 1000 ppm =1 g/L= 1000 µg/ml
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.