Roll Bag Dispensers, Eagle MHC
Fournisseur: Eagle Group
Certificats
A propos de cet article
Dispenses any product on tube stock greater than 1" I.D. Stainless steel or chrome finish available.
- Consists of four posts grooved on 1" centers for adjustability
- Two 14" x 48" (356 x 1219mm) wire shelves
- Eight removable dispenser bars with 40" (1016mm) length
- One three sided channel frame
- Two 30" deep outrigger bases for stability
An effective means of dispensing products on rolls or spools. An easy way to establish protocol by color of cleanroom waste, avoiding inadvertent mixing of non-compatible items. Allows significant inventory to be stored at the point of use.
Customers Who Bought This Also Bought