Nexion ICP-MS Calibration Standard for Set Up 8, VeriSpec®, Ricca Chemical Company
Fournisseur: Ricca Chemical
Certificats
A propos de cet article
8 elements.
1 ppb: Be, Ce, Fe, In, Li, Mg, Pb, U
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.