CORIO™ CP Immersion Heating Circulators, JULABO USA, Inc.
Fournisseur: JULABO
Certificats
A propos de cet article
CORIO™ CP immersion heating circulator with adjustable clamp for internal temperature applications to 50 L from 20 to 200 °C.
- Infinitely adjustable pressure pump
- Bright LED display
- USB and RS232 ports
For internal temperature applications. Clamps easily on bath tanks with walls to 30 mm thick. Immersion depth: 7.5 to 16.5 cm.
CORIO CP circulator features a bright LED display button controls. The USB and RS232 ports facilitate external communication to PC or PLC systems. Pump connections: M16×1 male threads.
Certifications: CE certified.
Delivery: Includes CORIO CP immersion circulator and adjustable clamp. Supplied without accessories.
Customers Who Bought This Also Bought