OIC Strong Binding Self-Adhesive Fasteners, Eaton Office Supply
# de catalogue 470152-504
Fournisseur: EATON OFFICE SUPPLY
Spécifications
- Description:Strong Binding Self-Adhesive Fasteners, Steel, Silver, 100 per Box
- Size:1" Size
- Cat. no.:470152-504
- Dimensions:2.8" L
- Supplier No.:OIC99857
Spécifications
A propos de cet article
Self-adhesive fasteners attach punched documents to any file folder or filing surface.
- Strong binding self-adhesive fasteners
Strong adhesive keeps fastener permanently attached. Fasteners are crafted from the same corrosion-resistant, tempered steel as standard fasteners. Capacity is 1".
Customers Who Bought This Also Bought