Scotch® Tape, Gloss Finish Transparent, Acrylate, 1" Core, Eaton Office Supply
# de catalogue 470201-404
Fournisseur: EATON OFFICE SUPPLY
Spécifications
- Dimensions:0.50W×37.50"L
- Description:Gloss Finish Transparent Tape
- Cat. no.:470201-404
- Supplier No.:MMM-144A
Spécifications
A propos de cet article
Scotch® Transparent Office Tape is a glossy tape that is crystal-clear for a professional appearance.
- Glossy
- Non-yellowing
- Photo-safe
- Transparent
- Compact handheld dispenser
Instant adhesion delivers excellent holding power for your most critical taping tasks. Clear when applied, it doesn't yellow with aging. Tape pulls off the roll smoothly and cuts easily. It is ideal for multipurpose sealing, label protection, general-purpose wrapping and mending. It is also photo-safe (determined in accordance with ISO Standard 18916). The tape comes on a compact handheld dispenser that stores easily in a drawer.
Customers Who Bought This Also Bought