Scotch® Foam Mounting Squares, Eaton Office supply
# de catalogue 470150-138
Fournisseur: EATON OFFICE SUPPLY
Spécifications
- Dimensions:1W×1"L
- Description:Permanent Mounting, 16 per Pack
- Cat. no.:470150-138
- Supplier No.:MMM 111
Spécifications
A propos de cet article
Double-coated foam squares are ideal for mounting artwork and decorations to a variety of surfaces.
- Mounting artwork
- Double-sided
- Permanent mounting
- Adhesive
General-purpose, heavy-duty foam tape is designed for mounting light to medium-weight objects to both walls and other surfaces. Each mounting square conforms to many textured walls and has adhesive on both sides. Adhesive is permanent. Each square can hold up to 2 lb.
Customers Who Bought This Also Bought