Fellowes Slim Jewel Cases, Book Fold, Eaton Office Supply
# de catalogue 470150-048
Fournisseur: EATON OFFICE SUPPLY
Spécifications
- Color:Clear, Black
- Description:CD Jewel Case, 100 per Pack
- Cat. no.:470150-048
- Dimensions:5.6×4.9"
- Supplier No.:FEL-98335
Spécifications
A propos de cet article
Durable, slim replacement cases hold CDs/DVDs in half the space of a standard jewel case and include an area for an insert booklet.
- Each jewel case stores one CD/DVD and booklet
- Cases are made of plastic
- 100% recyclable
Cases are made of plastic. Each jewel case stores one CD/DVD and booklet. Protective ports and spines prevent damage to CDs/DVDs. Jewel cases are 100 percent recyclable
Customers Who Bought This Also Bought