Removable ID Labels, Avery
Catalog # 470094-286
Supplier: EATON OFFICE SUPPLY
Specifications
- For Use With:Laser or inkjet printer
- Description:Lables, removable package, 840 per pack
- Dimensions:1.75×0.5"
- Cat. no.:470094-286
- Supplier No.:5422
Specifications
About this item
Self-adhesive labels are perfect for addressing, identifying, updating, organizing and price marking.
- Labels stick, stay and remove cleanly for easy updates
- Multipurpose labels adhere to smooth surfaces
- Laser or inkjet printer
Multipurpose labels adhere to smooth surfaces including paper, cardboard, plastic, wood, glass and metal. Labels come on a 4×6" sheet size. Write on or print using your laser or inkjet printer.
Ordering information: Accessories not included.
Customers Who Bought This Also Bought