Hei-VAP Ultimate Control, Rotary Evaporators, Heidolph®
Fournisseur: Heidolph NA, LLC
Certificats
A propos de cet article
Designed as precise setting of rotation speed and heating bath temperature on the digital touch display or for quick access during an active process, directly via the knobs on the operating panel.
- Clear display of all values
- Central operating panel with 7" touch display
- With timer, limitation of maximum temperature, multilingual menu and text display for error messages
- LED ring lights and pictogram to display the activity even from a distance
- USB and Micro SD plus LAN / RS 232 interfaces
- Optional display of vapor temperature
It has integrated solvent library to facilitate adjustment of the required vacuum on an external vacuum controller with timer, limitation of maximum temperature, multilingual menu and text display for error messages. Available in Hand Lift (HL) and Motor Lift (ML) Configurations.
Ordering information: G1 glassware set is a diagonal coil condenser. G3 glassware set is a vertical coil condenser. G3 XL glassware is a higher evaporation rate condensor. Glassware set G5 is a dry ice condenser. Glassware set G6 is a vertical condenser.
Spécifications
- Catalog No:
- 76338-460
- 76338-458
- 76338-440
- 76338-452
- 76338-474
- 76338-450
- 76338-472
- 76338-456
- 76338-478
- 76338-454
- 76338-476
- 76338-444
- 76338-466
- 76338-442
- 76338-464
- 76338-448
- 76338-470
- 76338-446
- 76338-468
- 76338-462
- Model:
- Hei-VAP Ultimate, Control ML, G1
- Hei-VAP Ultimate, Control HL, G6 B
- Hei-VAP Ultimate, Control HL, G1
- Hei-VAP Ultimate, Control HL, G 3B
- Hei-VAP Ultimate, Control ML, G3 B XL
- Hei-VAP Ultimate, Control HL, G1B
- Hei-VAP Ultimate, Control ML, G3 B
- Hei-VAP Ultimate, Control HL, G5 B
- Hei-VAP Ultimate, Control ML, G6 B
- Hei-VAP Ultimate, Control HL, G 3B XL
- Hei-VAP Ultimate, Control ML, G5 B
- Hei-VAP Ultimate, Control HL, G3 XL
- Hei-VAP Ultimate, Control ML, G5
- Hei-VAP Ultimate, Control HL, G3
- Hei-VAP Ultimate, Control ML, G3 XL
- Hei-VAP Ultimate, Control HL, G6
- Hei-VAP Ultimate, Control ML, G1B
- Hei-VAP Ultimate, Control HL, G5
- Hei-VAP Ultimate, Control ML, G6
- Hei-VAP Ultimate, Control ML, G3
- Rotation speed:
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- 10 - 280 rpm
- Bath capacity:
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- 4.5 L
- Temperature range:
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- 20…210 °C
- Max. flask capacity:
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- 5 l
- Bath Ø:
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- 255 mm
- Accuracy:
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C
- ±1 °C