Statfree Z2™ Dissipative 3-Layer Vinyl Mat, Desco
Fournisseur: Desco
Certificats
A propos de cet article
Statfree Z2™ Dissipative 3-Layer Vinyl Mats provide reliable path to ground and eliminate static charge.
- Heat fused 3-layer vinyl composite
- Dissipative (1×10E6 < 1×10E9 ohms Rtt) abrasion resistant top layer, with limited lifetime warranty
- Conductive (<10E4 Rtt) mesh middle layer bottom layer, with limited lifetime warranty
- Dissipative (1 x 10E6 < 1×10E9 ohms Rtt) cushioned bottom layer, with limited lifetime warranty
- Made in the United States of America
Eliminates static charge generation as a worksurface table mat, shelf liner, and cart liner. Electricals will not drift for the life of the mat. The dedicated conductive layer reduces the resistance to ground (Rtg) from the top of the surface to the groundable point ground, and allows the mats to be used with monitors.
Cushioned bottom layer provides additional energy absorption for components susceptible to physical shock.
Certifications: Lead-free, RoHS compliant.
Ordering information: Vinyl Mat includes mat, ground cord, universal snap kit, and Reztore™ surface and mat cleaner. Vinyl Roll includes mat roll only.
Customers Who Bought This Also Bought