ESD Workstation Cover, Desco
Supplier: Desco
Certificates
About this item
ESD Workstation Cover protects the workstation from ESD, dust, and other contaminants.
- Groundable workstation cover
- Surface resistance: 1×10E5 < 1×10E7 Rtt
- Minimum of 9% carbon conductive nylon fibers with bound edges
- 10 mm (3/8") grounding socket
- Made in the United States of America
Cover attenuates electrostatic fields and electrostatic discharges (ESD). Minimum of 9% carbon conductive nylon fibers with bound edges provide continuous and consistent charge dissipation throughout lifetime of cover.
Grounding socket (10mm) allows cover to be grounded to the workstation’s common point ground.
Customers Who Bought This Also Bought