Reztore® Antistatic Coating, Desco
# de catalogue 89131-018
Fournisseur: Desco
Product available on GSA Advantage®
Spécifications
- Volume:1.1 L
- Description:Reztore® Antistatic Coating
- Cat. no.:89131-018
- Supplier No.:10415
Spécifications
A propos de cet article
Reztore® Antistatic Coating keeps triboelectric charges well below 200 volts per EIA 625.
- Long lasting hard coat formula
- Static dissipative
- Non-toxic
- Removes contaminating particulate
- Rtt: 10E6 ohms to < 1×10E9 ohms per ANSI/ESD S4.1
- Bag-in-box packaging available
- Lead-free RoHS compliant
- Made in The United States of America
Long lasting hard coat formula protection lasts three times as long as other leading brands. Non-toxic and safe to use coating provides a safe path to ground. Removes contaminating particulates and eliminates possible ESD event. This product has been certified to meet the RoHS requirement.
Customers Who Bought This Also Bought