VWR® Blue Twist-Seal Bags
# de catalogue 76264-506
Fournisseur: VWR International
Spécifications
- Capacity:650 ml (22 oz.)
- W×L:140×229 mm (5.5×9")
- Description:PE blue sterile bags
- Thickness:3 mil
- Cat. no.:76264-506
Spécifications
A propos de cet article
These blue twist-seal printed bags with safety tab and 2 round wires are made of virgin PE and have been specifically designed for the food industry.
- Blue color tear-off for increased visibility
- Distinct color to avoid possible foreign matter contamination
- Visual recognition of the product, but can also be used to segregate within a factory environment
- Optimized opening area to facilitate insertion of sample
- Lot number traceability
- All sterile sampling bags are also free from RNase, DNase and pyrogens
The bags accommodate solid, semi-solid and liquid compounds.
Certifications: Food contact approved.
Customers Who Bought This Also Bought