ContactUsLinkComponent
Practi-Fentanyl™ 5 ml Ampule for Training, Wallcur
Practi-Fentanyl™ 5 ml Ampule for Training, Wallcur
# de catalogue 470232-940
Fournisseur:  WALLCUR, LLC.
Practi-Fentanyl* 5 mL Ample
Practi-Fentanyl™ 5 ml Ampule for Training, Wallcur
# de catalogue 470232-940
Fournisseur:  WALLCUR, LLC.
Spécifications
  • Capacity:
    5 ml
  • Description:
    Traning Fentanyl Ampules, 5 ml
  • Cat. no.:
    470232-940
Spécifications
A propos de cet article
Medication admistration simulation and training.
  • For training purposes only
Practi-Fentanyl™ 5 ml Ampule for clinical training, accurately simulates 250 mcg/5 ml (50 mcg/ml) (0.05 mg/ml) of the synthetic opioid analgesic, Fentanyl. Each of it comes as a labeled 5 ml ampule containing distilled water.
Caution: Not for human or animal use.
Customers Who Bought This Also Bought