Jiffy Peat Pellets, 36 mm, Plantation Products
# de catalogue 470205-660
Fournisseur: PLANTATION PRODUCTS, INC.
Spécifications
- Description:Peat Pellets
- Cat. no.:470205-660
- Supplier No.:5701
Spécifications
A propos de cet article
Jiffy peat pellets are perfect for starting your seeds indoors.
- 36 mm Ø
- Pellets are 100% biodegradable
They are perfect for starting seeds and cuttings. Just expand with water, plant your favorite seeds, and when ready, the whole pellet and plant can be planted directly in the ground or in a container.
Packaging: Bulk pack with 1000 pieces.
Customers Who Bought This Also Bought