ContactUsLinkComponent
undefined | Electrophoresis Stains
Catalog # 76204-684
Supplier:  Biotium
CAS Number:  
Catalog # 76204-684
Supplier:  Biotium
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Size:
    10 ml
  • Cat. no.:
    76204-684
Specifications
About this item
DNAzure® Blue Nucleic Acid Gel Stain is an ultrasensitive reagent for visible staining of dsDNA in agarose or polyacrylamide gels.
  • Deep blue bands visible by the naked eye following 5-30 min light exposure
  • Ultrasensitive detection, as little as ~1 ng DNA
  • Simplified DNA band excision, without the need for DNA-damaging UV light
  • Bands are stable for weeks after color development
  • Compatible with downstream applications such as sequencing and cloning
  • Expensive gel documentation systems not required for imaging
After incubation with DNAzure® Blue Nucleic Acid Gel Stain, the gel must be exposed to bright light to facilitate the development of dark blue DNA bands.
Customers Who Bought This Also Bought