ProPette™ LE Adjustable Pipettors, MTC Bio
Fournisseur: MTC BIO, INC.
Certificats
A propos de cet article
The ProPette LE™ is the next generation of high precision liquid handling instruments from MTC Bio. This addition to the Propette family features improved actuation forces and a nearly 20% reduction in overall weight.
- Adjustable with Roto-Lock™
- Light weight with reduced forces
- Full volume range (0.1 µl to 10 ml)
- Superior accuracy and reproducibility
- Outstanding user comfort
Every aspect of accuracy, reliability and user comfort has been carefully reviewed in designing these high precision pipettes. The result is an advanced pipette that exhibits superior accuracy and ergonomics and is comfortable to use, even during prolonged and repetitive pipetting.
Certifications: CE marked.
Delivery: Includes calibration key.
Customers Who Bought This Also Bought