Memmert Paraffin Ovens, UNpa Series
Fournisseur: Memmert
Certificats
A propos de cet article
Stainless steel interior, complete with manufacturers calibration certificate at 80 °C (measured at the center point of the chamber).
- Stainless steel interior
- Textured stainless steel exterior
- Nearly gas tight chamber - no feed or exhaust air
- Avoids danger of ignition of residue or damage to mechanical and electronic components
Reliable sample preparation for forensics, histology and microbiology. Five model sizes to chose from, sizes 32 to 161 litres volume. Twin Display units with high degree of thermal/heating safety with muliple overtemerature protection. No air inlet, no exhaust port - nearly gas tight chamber. Reliable sample preparation equipment that gaurantees precise, even warming of the embedding medium without loss of quality.
Certifications: CUL Approved.
Delivery: Supplied with two stainless steel grid shelves.
Customers Who Bought This Also Bought