Spécifications
- Length:0.75"
- Cat. no.:89134-232
- Gauge:20G
- Supplier No.:WHIS-20-75
Spécifications
A propos de cet article
These winged infusion sets are designed for easy insertion into subcutaneous Vascular Access Ports (VAPS), and can be used for continuous or bolus infusions.
- Huber needle allows easy penetration without coring
- Alternate Huber needle configurations are available
- Connect to standard syringes or a luered fitting
- Customizable
Each set is comprised of a 90° Huber Needle with suture wings, connected to 10" of mirco-bore tubing, with a luer lock fitting. Supplied sterile.