ContactUsLinkComponent
Quick Connect™ Jacket Adaptors, SAI Infusion Technologies
Quick Connect™ Jacket Adaptors, SAI Infusion Technologies
Catalog # 89134-212
QUICK CONNECT JACKET ADAPTER
Quick Connect™ Jacket Adaptors, SAI Infusion Technologies
Catalog # 89134-212
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Quick Connect™ jacket adaptor with injection cap, 23 G pin
  • Cat. no.:
    89134-212
  • Supplier No.:
    QCJA-23
Specifications
About this item
The Quick Connect™ Jacket Adaptor is designed for researchers who prefer a woven rodent jacket over an elastomeric harness for continuous or intermittent infusion. The Jacket Adaptor utilizes the same luer design as the Quick Connect™ Harness, but connects securely to a cloth jacket with Velcro™, and can be adjusted as the animal grows.
  • Attached injection cap can be removed, or pierced with a Huber needle
  • Choose from two catheter connector pin sizes to fit 21G and 23G catheters
  • To be used with an SAI/Lomir Jacket
  • Fit can be adjusted as the animal grows
  • Individually packaged
  • EtO exposed
These jackets are part of SAI's Quick Connect™ System, which includes the Quick Connect™ Tether, Harness, and Swivel for tethered small animal infusion studies.
Packaging: Packaged individually and sterile.
Customers Who Bought This Also Bought