ContactUsLinkComponent
10 L Jacketed Filter Reactor Systems, Air Motor, Chemglass
10 L Jacketed Filter Reactor Systems, Air Motor, Chemglass
# de catalogue 89209-342
Fournisseur:  Chemglass
Small Business Enterprise
Process Reactor 10L Jackt Air
10 L Jacketed Filter Reactor Systems, Air Motor, Chemglass
# de catalogue 89209-342
Fournisseur:  Chemglass
Small Business Enterprise
Spécifications
  • Description:
    Jacketed Cylindrical Process Filter Reactor System, Complete
  • Motor Type:
    Air motor
  • Cat. no.:
    89209-342
  • Supplier No.:
    CG-1958-F611
Spécifications
A propos de cet article
Complete reactor system on open air, autocenter stand.
Maximum temperature limit 150 °C for filter base.
Customers Who Bought This Also Bought