Condensers, 'C' Assembly, Constant Feed, for Rotary Evaporators, Plastic Coated, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Two piece cold trap style Condenser for use with rotary evaporators.
- Condenser is supplied complete with all fittings necessary to connect the continuous feed feature
- Along with Chemglass new style PTFE centering ring which makes a vacuum tight seal between the inner cold finger and the plastic coated outer sleeve
- Also supplied with a UHMW lid with knob
Condenser has an additional 19/38 outer joint sealed onto the condenser to allow continuous feed of volumes exceeding the capacity of the evaporating flask. The 19/38 joint is sealed at an angle which allows the condensate to fall into the receiving flask without running down the addition tube into the evaporating flask. This continuous feed feature eliminates the need for a separate filtration step to remove drying agents or other particulate matter from organic solutions prior to rotary evaporation. The ferrule style connection eliminates O-ring connections, which are prone to degradation with long term exposure to organic solvents.
Customers Who Bought This Also Bought