Drying Tubes, 75°, Chemglass
Fournisseur: Chemglass
Certificats
Certificats
A propos de cet article
Drying tubes feature the 75° angle drying tube.
- These for use in fume hoods where space is at a minimum
- Complete with rubber stopper, glass inlet tube
Drying tube is designed with a lower inner joint.
Customers Who Bought This Also Bought
PYREX® Disposable Culture Tubes, Rimless, Dispenser Pack and Bulk Pack, Corning
Fournisseur: Corning
$483.39
Sample Vials, Borosilicate Glass, WHEATON®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$411.14
Kimble® Disposable Pasteur Pipettes, Soda Lime, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$286.15
Shorty Vials®, Borosilicate Glass, Wheaton, DWK Life Sciences
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
$236.16
Filtration, Microanalysis Vacuum Filter Holder Set, 25 mm with Stainless Filter Support, United Scientific Supplies
Fournisseur: United Scientific Supplies
$88.50
EP Scientific Silanized Culture Tubes and Vials, Thermo Scientific
Fournisseur: Thermo Scientific
$1,853.13
KIMBLE® RAY-SORB® Amber GL 45 Media Bottle, DWK Life Sciences
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
$128.55
ACCUFORM® SSR™ Vials and Black Phenolic Screw Caps, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
$982.38 to $3,929.51