Spécifications
- Volume:1 ml
- Description:Isotonic Saline (NaCl) Pre-filled Syringe with Attached Cap
- Sterility:Non sterile
- Cat. no.:89134-214
- Packaging:10/Pack, 10 Packs/Box
- Supplier No.:SC1-1
Spécifications
A propos de cet article
Prefilled syringes for flushing an indwelling catheter or port system. This prefilled option can saves time, space, and money.
- Saline (0.9%) filled syringe, customization available
- Syringe sizes from 1 to 60 ml
- Volumes from 0.2 ml to filled
- Syringes with caps, or with blunt needles already attached
No need to unwrap individual syringes, no need to store physiologic containers, no need to formulate solutions, and no need to spend valuable time filling syringes.
Sterile solution and fluid path. Exterior surface of syringe is non sterile.
Heparinized saline syringes and catheter maintenance kits are also available.