ContactUsLinkComponent
Filter Reactor Support Bases, Chemglass
Filter Reactor Support Bases, Chemglass
Catalog # 10170-640
Supplier:  Chemglass
Small Business Enterprise
Filter Support Assembly, Complete Without Filter, 200mm
Filter Reactor Support Bases, Chemglass
Catalog # 10170-640
Supplier:  Chemglass
Small Business Enterprise
Specifications
  • Description:
    Filter Support Assembly, 200 mm, Complete without Filter
  • Cat. no.:
    10170-640
  • Supplier No.:
    CG-1959-F200
Specifications
About this item
Filter reactor support bases.
  • Maximum temperature: 150 °C
Complete filter reactor support bases with temperature limit for filter base.
Customers Who Bought This Also Bought