Adapters, Distilling/Splitting, Process Reactors, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Compact and simple distilling/splitter adapter for use with process reactors.
- Control valve is 0 to 4 mm PTFE
- A 1/2" O.Ø distillate takeoff for use with compression fittings and TFE tubing
- Plugs are supplied complete with Perfluoro O-rings
Vapor flows upward freely through the large vapor holes to the condenser. Distillate flow is directed using the valve to either the receiving flask or back to the reactor.
Customers Who Bought This Also Bought