Funnels, Heavy Duty, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Heavy duty funnels are autoclavable.
- Highly recommended for continuous laboratory work
- Steam autoclavable at 121 °C (250 °F)
Sturdy polypropylene funnels with ridges to prevent vapor lock.
Customers Who Bought This Also Bought