ContactUsLinkComponent
Multi-Directional Swivel Mount Assembly, SAI Infusion Technologies
Catalogue # 89134-254
undefined
Multi-Directional Swivel Mount Assembly, SAI Infusion Technologies
Catalogue # 89134-254
Supplier Number:  GMSHK
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Multi-Directional Swivel Mount Assembly, with Gimbal Mount and Adjustable 8" Post
  • Cat. no.:
    89134-254
  • Supplier No.:
    GMSHK
Spécifications
À propos de cet article
Completely adjustable, multi-directional swivel mounting system made of anodized aluminum.
  • Reduce animal stress and tether tangles with the gimbal mount's unparalleled 4-axes of movement
  • Stainless steel base quickly attaches to any style caging and is cage washer-resistant
  • Quick-release swivel holder allows for continuous infusion during body weights and cage changes
  • Durable anodized aluminum components
The swivel is easily raised and lowered with a thumb screw, and the 8" post can be trimmed to any height. Interchangeable fittings allow use of the multi-directional swivel holder with or without the counter balance arm.
Caution: This product is for laboratory animal use only.