Jacketed Filter Reactor Systems, 20 L, Air Motor, Chemglass
# de catalogue 89209-344
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Jacketed Cylindrical Process Filter Reactor System Complete
- Motor Type:Air motor
- Cat. no.:89209-344
- Supplier No.:CG-1958-F621
Spécifications
A propos de cet article
Jacketed filter glass reactors feature autocenter stand.
- Complete reactor system on open air, autocenter stand
- 150 °C maximum temperature limit for filter base
- Filter base eliminates material transfers and significantly reduces losses
Jacketed filter glass reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis.
Customers Who Bought This Also Bought