Distilling Adapters, Process Reactors, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
High quality distillation Adapters are for use with flasks or reaction vessels.
- Adapter has two top outer joints and a lower inner
- Adapter has a lower inner joint and two top outer joints
Similar to CG-1235, but with the lower stopcock replaced with a 0 to 14 mm treaded valve having Perfluoro O-rings and a 1/2" O.D. glass distillate takeoff for use with PFA compression tubing fittings. Style “B” with lower angled joints are designed for use with flasks or reaction vessel lids that have a 10° or 20° angled side neck.
Customers Who Bought This Also Bought