Reaction Vessel Lids, 5-Necks, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Reaction vessel lids feature 30° angle side necks.
- Lid has a flat ground surface
- Allow flat ground surface to be greased or used with an O-ring
Five neck vessel lid to fit CG-1920 through CG-1936 vessels.
Customers Who Bought This Also Bought