Heating Mantles, Lower Portions, Unjacketed Cylindrical Vessels, Chemglass
Supplier: Chemglass
Certificates
About this item
For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels.
- TM Series aluminum housed glass fabric heating mantle
- With hole centered in the bottom
Mantle is 115 volts and is supplied complete with a separable 4 feet 2-wire cord and locking connector.
Customers Who Bought This Also Bought