Adapter, Flow Control, Standard Taper Joints, Chemglass
# de catalogue 80066-750
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- Description:Adapter, Flow Control, Chromatography
- Cat. no.:80066-750
- SJ Joint No.:45/50
- Supplier No.:CG-1191-02
Spécifications
A propos de cet article
Flow control adapter for use with CG-1188 and CG-1189 chromatography columns along with CG-1190 reservoirs.
- Adapter has a 0-4 mm Chem-Cap® metering plug
Customers Who Bought This Also Bought