Drying Tubes, Straight, Top and Outer Joints, Chemglass
# de catalogue 80067-982
Fournisseur: Chemglass
Small Business Enterprise
Spécifications
- ST Joint No.:14/20
- Description:Drying Tube, Straight
- Cat. no.:80067-982
- Supplier No.:CG-1294-14
Spécifications
A propos de cet article
Drying tubes feature straight, top and outer joints.
- Top standard taper outer joint
- Same size as the lower inner joint
Similar to CG-1293 but with the addition of a top standard taper outer joint of the same size as the lower inner joint.
Customers Who Bought This Also Bought