Flowmeters, Bubble, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Flowmeter used for the control of carrier gasses.
- Projection at top breaks bubbles
Hose connection has a outer diameter of 9 mm at the middle ring.
Delivery: Supplied complete with rubber bulb.
Customers Who Bought This Also Bought