Order Entry
ContactUsLinkComponent
Human Recombinant BMP-4 (from HeLa cells)
Human Recombinant BMP-4 (from HeLa cells)
# de catalogue 76303-720
Fournisseur:  Peprotech
Numéro CAS:  
Human Recombinant BMP-4 (from HeLa cells)
# de catalogue 76303-720
Fournisseur:  Peprotech
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Animal-free:
    Y
  • Gene ID:
    652
  • Protein Function:
    Cytokines and Growth Factors
  • Protein/peptide type:
    Recombinant
  • Source:
    HeLa cells
  • Species:
    Human
  • Conjugation:
    Unconjugated
  • Size:
    50 µg
  • Biological activity:
    Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 2-8 ng/ml.
  • Protein synonyms:
    DVR4|Bone Morphogenetic Protein-4|BMP-2B
  • Protein/peptide name:
    BMP-4
  • Purity:
    97%
  • Molecular weight:
    25.6 kDa
  • Sequence:
    HHSQRARKKN KNCRRHSLYV DFSDVGWNDW IVAPPGYQAF YCHGDCPFPL ADHLNSTNHA IVQTLVNSVN SSIPKACCVP TELSAISMLY LDEYDKVVLK NYQEMVVEGC GCR
  • Endotoxin level:
    low
  • Formulation:
    Lyophilized
  • Shipping temperature:
    Ambient
  • Cat. no.:
    76303-720

Spécifications

A propos de cet article

Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-beta superfamily of structurally related signaling proteins. Members of this superfamily are widely distributed throughout the body, and are involved in diverse physiological processes during both pre- and postnatal life. Like BMP-7, BMP-4 is involved in the development and maintenance of bone and cartilage. Reduced expression of BMP-4 is associated with a number of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Recombinant Human BMP-4, expressed in HeLa cells, is a 25.6 kDa homodimeric glycoprotein.

Customers Who Bought This Also Bought