To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Secondary
- Antigen name:IP-10 (CXCL10)
- Clonality:Polyclonal
- Gene ID:15945
- Host:Rabbit
- Reactivity:Mouse
- Antigen symbol:IgG
- Conjugation:Biotin
- ELISA:Yes
- Size:1 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:gamma-Interferon Inducible Protein-10, CXCL10, crg-2
- Storage buffer:See Product Data Sheet for more information.
- Molecular weight:8.7 kDa
- Storage temperature:Store at -20°C prior to reconstitution.
- Shipping temperature:Ambient
- Immunogen:E.coli-derived Recombinant Murine IP-10 (CXCL10) (PeproTech catalog# 250-16)
- Purification:Affinity purified
- Packaging:PP Vial
- Supplier No.:500P129BT1MG
Spécifications
A propos de cet article
Produced from sera of rabbits immunized with highly pure Recombinant Murine IP-10 (CXCL10). Anti-Murine IP-10 (CXCL10) specific antibody was purified by affinity chromatography and then biotinylated.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Murine