To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:IL-4
- Clonality:Polyclonal
- Gene ID:16189
- Host:Rabbit
- Reactivity:Mouse
- Antigen symbol:IgG
- Conjugation:Unconjugated
- ELISA:Yes
- Size:50 ug
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:BCGF, BCDF, B cell stimulating factor (BSF-1)
- Storage buffer:See Product Data Sheet for more information.
- Molecular weight:13.5 kDa
- Storage temperature:Store at -20°C prior to reconstitution.
- Shipping temperature:Ambient
- Immunogen:E.coli-derived Recombinant Murine IL-4 (PeproTech catalog# 214-14)
- Purification:Affinity purified
- Packaging:PP Vial
- Supplier No.:500P5450UG
Spécifications
A propos de cet article
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-4. Anti-Murine IL-4-specific antibody was purified by affinity chromatography employing an immobilized Murine IL-4 matrix.
Type: Primary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Murine