CleanTex™ ClearView Wipes, Advantus
# de catalogue 58019-502
Fournisseur: Advantus
Spécifications
- W×L:7.6×10.2 cm (3×4")
- Description:ClearView Wipes
- Cat. no.:58019-502
- Packaging:80/Box
- Supplier No.:CT803
Spécifications
A propos de cet article
Lint-free wipes made of 100% polyester allow users to clean screens and monitors without streaking.
- The original screen cleaner, designed to clean and control static on large, heavily soiled screen surfaces
- Use Hydrosorb I lint-free wiper to wipe screen dry to avoid streaking
- Static control formula reduces static buildup
- Not for use on polarized anti-glare screens
These wipes have ultra-saturated applicator made of 100% polyester which withstands vigorous cleaning.
Customers Who Bought This Also Bought