CleanTex™ Tex Stat Wipes, Advantus
# de catalogue 58019-516
Fournisseur: Advantus
Spécifications
- Description:Tex Stat Wipes
- L×W:12.7×17.8 cm (5×7")
- Cat. no.:58019-516
- Supplier No.:CT814
Spécifications
A propos de cet article
Cleans dirt and dust on office equipment and workstations without causing static buildup.
- Eliminates static
- Presaturated pad with antistatic and cleaning agents
- Cleans and control static on synthetic surfaces such as terminal housings, workstations, equipments, desktops, and media files
CleanTex™ Tex Stat Wipes are multi-surface pads suitable for cleaning dirt and dust on office equipment and workstation. Each pack is supplied with 72 pieces, assuring a long service after purchase.
Customers Who Bought This Also Bought