CleanTex™ Hydrosorb I Polyester/Cellulose Non-Woven Wipes, Advantus
Fournisseur: Advantus
Certificats
A propos de cet article
Combines the highly absorbent properties of a natural fiber with the cleanliness and strength of a synthetic wipe to clean up aqueous spills, precision components, and lab equipment.
- Hydrosorb I wipes are fabricated from a hydroentangled, non-woven blend of 55% cellulose and 45% polyester
- CleanTex™ Hydrosorb I wipes offer excellent strength, both wet and dry
- Compatible with most solvents
- No chemical binders or additives
- Low particle counts and extractable levels make these wipes virtually lint-free
- Autoclavable
These CleanTex™ Hydrosorb I Wipes for superior durability and absorbency. Hydrosorb I is ideal for general wiping and spill control in controlled environments. Designed for general-purpose wiping where economy and cleanliness are of critical importance. These wipes are ideal for applications where high absorbency and low extractable levels are of paramount concern.
Customers Who Bought This Also Bought