SST Series Heat-Tolerant Brushes, Conductive, Gordon Brush
Fournisseur: Gordon Brush
Certificats
A propos de cet article
Stainless steel applicator brushes with a lightweight stainless steel handle.
- Stainless steel handle is chemical resistant and non-magnetic
- Maximum use temperature of 900°F for short term exposures
Manufactured using strong mechanical clamping for maximum fiber retention, and is adhesive-free to prevent contamination on surface areas caused by leaching.
Customers Who Bought This Also Bought