A propos de cet article
This DetachKit is used for gentle and effective detachment of adherent primary human cells in routine subculturing.
- Each batch is tested on primary human cells
- Can be used for primary animal cells and cell lines
- Comes with a Trypsin/EDTA ratio of 0.04%/0.03%
Each DetachKit consists of three components: HEPES BSS (HEPES buffered balanced salt solution), Trypsin/EDTA solution and TNS (Trypsin Neutralization Solution).
HEPES BSS contains 30 mM HEPES, D-Glucose, NaCl, KCl, Na-Phosphate and Phenol Red. TNS contains 0.05% Trypsin Inhibitor from soybean and 0.1% Bovine Serum Albumin.