Young's Modules of Wires Apparatus, Eisco Scientific
# de catalogue 470159-556
Fournisseur: Wards
Spécifications
- Description:Young's modules of wires apparatus
- Cat. no.:470159-556
- Supplier No.:PH0328A
Spécifications
A propos de cet article
Learn how to measure the stiffness of wires.
- Aluminum cast frame includes arm with bubble level
- Micrometer screw
- Iron weight (approximate): 2.6 lbs.
The Young's modulus of wires apparatus provides all of the necessary tools to study the concept of the measure of the stiffness of a material (Young's Modulus). It provides the tools to study the stress and strain on a material in a visually and kinesthetically effective manner.
Delivery: Includes aluminum cast frame, micrometer screw, wire holder, two screws, iron weight with hook, and two wires.
Customers Who Bought This Also Bought