Round Flash Lamp Bulbs, 6 V, Eisco Scientific
# de catalogue 470230-380
Fournisseur: Wards
Spécifications
- Voltage:6 V
- Description:Round Flash Lamp Bulbs
- Cat. no.:470230-380
Spécifications
A propos de cet article
Suitable for flash lamps, spotlights and panel lamps.
- Size E10 screw base
- For use with basic circuit kits
- Works with flash lamps, spot lamps and panel lamps
- 6.0 V with M.E.S. cap
Packaging: Supplied in pack of 10.
Customers Who Bought This Also Bought