Corepoint Scientific™ Controlled Room Temperature (CRT) Climatic Chambers, Horizon Scientific
Supplier: Horizon Scientific
Certificates
About this item
These CRT cabinets provide controlled room temperature environments for storing sensitive substances that require precise temperature management.
- Integrated temperature alarms
- Keyed door locks
- Forced draft air circulation
- Probe access
- Precise microprocessor temperature control
- Natural hydrocarbon refrigerants
- ENERGY STAR certified*
These specialized cabinets are ideal for scientific and medical applications. Both built-in and freestanding configurations are available. Built-in units are designed with front-facing ventilation systems, allowing easy installation into a bench or countertop. Freestanding units require a 4-inch margin on all sides for adequate ventilation. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76336-666, 76336-682, 76336-684, 76336-686, 76336-688, 76336-690, 76336-692, 76336-694, 76336-696, 76336-698, 76336-700, 76336-702, and 76336-704
Ordering information: All units have a two-year parts and labor warranty. Selected models come with an additional three-year compressor parts warranty. (Please refer to unit datasheets to confirm).
Specifications
- Catalog No:
- 76336-694
- 76336-696
- 76336-692
- 76336-702
- 76336-682
- 76336-704
- 76336-698
- 76336-700
- 76336-666
- 76336-688
- 76336-690
- 76336-684
- 76336-686
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants
- Natural Refrigerants