Squishy Circuits, Deluxe Kit
# de catalogue 470317-844
Fournisseur: SQUISHY CIRCUITS STORE LLC SE
Spécifications
- Description:Squishy Circuits Deluxe Kit
- Cat. no.:470317-844
Spécifications
A propos de cet article
The deluxe kit is our most complete Squishy Circuits kit.
Start circuit building right away with the deluxe kit, our most complete Squishy Circuits kit. With more items than the standard kit and with pre-made doughs, you can start creating and playing without all the unnecessary waiting! This kit also includes a deluxe battery holder which has a knob that allows your lights to blink and buzzers to beep at different speeds.
Delivery: Kit includes 1×Deluxe Battery Holder, 1 each - Piezoelectric Buzzer, Motor with Fan, Mechanical Buzzer, Switch and Rolling Pin, 40×LEDs (5 each or Red, Green, White, Yellow, Blue, Purple, Orange, and Color Changing), 1 Set of Dough Sculpting Tools, 1×7-Ounce Container of White Insulating Dough, and 7×7-Ounce Containers of Conductive Dough (in Red, Blue, and Green, Yellow, Orange, Purple, and Black).
Customers Who Bought This Also Bought