Hematite (Specular) Slab
# de catalogue 470236-112
Fournisseur: ADAM MOORE MO
Spécifications
- Description:Specular Hematite Slab
- Cat. no.:470236-112
Spécifications
A propos de cet article
Beautifully polished micaceous iron ore.
Average size: 3×4" to 5×6"
Customers Who Bought This Also Bought